Learn More
Medchemexpress LLC Sulfo-Cy3 NHS ester | 146368-16-3 | >95.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYD0823
Encompass_Preferred
Cy3-SE is a sulfonated cyanine dye-derived fluorescent labeling reagent with an excitation wavelength of approximately 515 nm and an emission wavelength of approximately 568 nm. It can interact with the π-π stacking of biomolecules (e.g., nucleoside monophosphates, proteins), inhibiting the photoisomerization process and increasing the fluorescence quantum yield and lifetime.
  • Sulfonated cyanine dye-derived fluorescent labeling reagent.
  • Excitation wavelength of about 515 nm and emission wavelength of about 568 nm.
  • Interacts with π-π stacking of biomolecules (e.g., nucleoside monophosphates, proteins).
  • Inhibits photoisomerization process.
  • Increases fluorescence quantum yield and lifetime.
  • Better enhancement effect on purines (dAMP, dGMP) than on pyrimidines (dCMP, dTMP) in the presence of nucleoside monophosphates (dNMPs).
Catalog No. 50-004-13559