Learn More
Medchemexpress LLC Sulfo-Cy5 amine | 2183440-44-8 | 98.0% | 740.97 | C38H52N4O7S2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD137510MG
Encompass_Preferred
Sulfo-Cy5 amine is a water-soluble, near-infrared fluorescent dye containing a primary amine for covalent conjugation to electrophilic targets. Sulfonate groups increase aqueous solubility, making the dye suitable for labeling biomolecules in biological buffers and imaging applications.
  • Water-soluble cyanine dye suitable for near-infrared imaging.
  • Primary amine functional group enables amine-reactive conjugation.
  • High purity (98.0%) supports reproducible labeling results.
  • Sulfonate groups enhance solubility in aqueous media and biological buffers.
  • Available in small milligram pack sizes for research-scale use.
Catalog No. 50-002-97847