Learn More
Medchemexpress LLC Sulfo-Cy5 azide | 1782950-80-4 | >90.0% | C37H48N6O10S3 | 10MG

Supplier: Medchemexpress LLC HY14574610MG
Sulfo-Cy5 azide is a water-soluble, azide-functionalized fluorescent dye intended for bioconjugation via click chemistry. It absorbs at 647 nm and emits at 663 nm, and is supplied as a blue powder with a molecular weight of approximately 833.01 g/mol and reported purity ≥90% (HPLC). The reagent is designed for labeling biomolecules in aqueous conditions and should be stored at -20 °C.
- Water-soluble for efficient labeling in aqueous buffers.
- Provides far-red fluorescence (λabs 647 nm, λem 663 nm) for imaging and detection.
- Azide functionality enables copper-catalyzed and strain-promoted click chemistry conjugation.
- High purity (>90.0%) suitable for sensitive applications.
- Blue powder form for easy aliquoting and storage.
- Stable when stored at -20 °C with a typical shelf life of 12 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.