Learn More
Medchemexpress LLC Sulfo-Cy5 azide | 1782950-80-4 | 98.97% | C37H48N6O10S3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY145746100MG
Encompass_Preferred
Sulfo-Cy5 azide is a near-infrared fluorescent probe that exhibits favorable click chemistry reactivity. It is used for fluorescence imaging, tissue and cellular visualization, and site-specific modification of antibodies. It can also be applied for RNA labeling and imaging and conjugated to targeting agents for fluorescence imaging in models of atherosclerosis and breast cancer.
  • Near-infrared fluorescent probe
  • Favorable click chemistry reactivity
  • Enables fluorescence imaging
  • Used for tissue and cellular visualization of PD-L1 in tumors
  • Allows site-specific modification of anti-PD-L1 antibodies
  • Applicable for RNA labeling and imaging
  • Can be conjugated to targeting agents for fluorescence imaging in atherosclerosis and breast cancer models
Catalog No. 50-003-54042