Learn More
Medchemexpress LLC Sulfo-Cy5 azide | 1782950-80-4 | 99.0% | C37H48N6O10S3 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY145746
Encompass_Preferred
Sulfo-Cy5 azide is a near-infrared fluorescent probe with favorable click chemistry reactivity. It is used for fluorescence imaging, enabling visualization of PD-L1 in tumors, site-specific modification of anti-PD-L1 antibodies, and RNA labeling. The probe can also be conjugated for imaging in atherosclerosis and breast cancer models.
  • Exhibits favorable click chemistry reactivity
  • Used for fluorescence imaging of PD-L1 in tumors
  • Facilitates site-specific modification of anti-PD-L1 antibodies
  • Applied in RNA labeling and imaging
  • Can be conjugated for imaging in atherosclerosis and breast cancer models
  • Detects ADP-ribosylation events via click chemistry
  • Conjugates to alkyne-functionalized TMV coat proteins (CuAAC)
Catalog No. 50-003-53281