Learn More
Medchemexpress LLC Sulfo-Cy5 hydrazide | 2055138-61-7 | 96.4% | 656.81 | C32H40N4O7S2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD188010MG
Encompass_Preferred
Sulfo-Cy5 hydrazide is a water-soluble, near-infrared fluorescent dye containing a hydrazide functional group for conjugation to aldehydes and ketones. It is optimized for imaging and fluorescence detection with excitation/emission maxima near 646/662 nm.
  • Near-infrared excitation and emission (646/662 nm).
  • Hydrazide reactive group for labeling aldehydes and ketones.
  • Water-soluble due to sulfonate groups, facilitating aqueous labeling reactions.
  • Solid form, light yellow to brown, high reported purity (96.39%).
  • Suitable for biomolecule labeling and fluorescence imaging applications.
Catalog No. 50-002-97190