Learn More
Medchemexpress LLC Sulfo-Cy7.5 DBCO | 99.8% | 1315.68 | C61H57K3N4O14S4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD224210MG
Encompass_Preferred
Sulfo-Cy7.5 DBCO is a sulfonated near-infrared fluorescent dye bearing a DBCO reactive group. The sulfonate groups increase water solubility, enabling aqueous labeling of biomolecules for imaging and tracking applications.
  • Water-soluble sulfonated cyanine dye suitable for aqueous labeling.
  • Near-infrared fluorescence for deep-tissue and low-background imaging.
  • DBCO reactive group enables copper-free click conjugation to azide-tagged biomolecules.
  • High purity suitable for sensitive protein and antibody labeling workflows.
  • Store dry at -20°C and protect from light and moisture.
Catalog No. 50-002-97463