Learn More
Medchemexpress LLC Sulfo-cyanine3 azide (potassium) | 1658416-54-6 | 95.3% | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD1274C10MG
Encompass_Preferred
Sulfo-cyanine3 azide (potassium) is a water-soluble, sulfonated cyanine fluorescent azide for bioorthogonal labeling via azide-alkyne click chemistry. It exhibits Cy3-like optical properties with excitation/emission near 548/563 nm, and is supplied as a potassium salt with a certificate of analysis for documented purity.
  • Cy3-like excitation/emission (548/563 nm).
  • Water-soluble sulfonated dye for aqueous labeling.
  • Reactive azide group for azide-alkyne click chemistry.
  • Supplied as a potassium salt with certificate of analysis and documented purity.
  • Suitable for labeling proteins, peptides, and oligonucleotides.
Catalog No. 50-002-97487