Learn More
Medchemexpress LLC Sulfo-cyanine5.5 | 210892-23-2 | 99.9% | 10 MG

Supplier: Medchemexpress LLC HYD092410MG
Cy5.5 (Sulfo-Cyanine5.5) is a near-infrared cyanine fluorescent dye used to label peptides, proteins, antibodies, and oligonucleotides for imaging and biodistribution studies. It is water-soluble, supplied as a purple to purplish-red solid, and exhibits strong absorption and emission in the near-infrared region (Ex 680 nm, Em 710 nm). Conjugation to biomolecules can be achieved using standard reactive derivatives.
- Near-infrared excitation/emission: Ex 680 nm, Em 710 nm.
- Water-soluble cyanine dye suitable for labeling biomolecules.
- Provided as a purple to purplish-red solid.
- Available in multiple pack sizes, including 10 MG and solution in DMSO.
- Storage: protect from light; in solvent: -80°C (up to 2 years), -20°C (up to 1 year).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.