Learn More
Medchemexpress LLC Sulfo-Cyanine7 NHS ester potassium | 1603861-95-5 | 90.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD135210MG
Encompass_Preferred
Sulfo-Cyanine7 NHS ester potassium is a water-soluble, amine-reactive N-hydroxysuccinimide (NHS) ester of a near-infrared cyanine dye supplied as the potassium salt. It is used to label primary amines on proteins, peptides, and other biomolecules to produce sulfo-Cyanine7-labeled conjugates for near-IR fluorescence imaging and related research applications.
  • Water-soluble near-infrared fluorescent dye.
  • Amine-reactive NHS ester for efficient conjugation to proteins.
  • Supplied as the potassium salt for enhanced solubility.
  • Suitable for labeling proteins, peptides, and other biomolecules.
  • Produces stable, near-IR fluorescent conjugates for imaging applications.
Catalog No. 50-002-97446