Learn More
Medchemexpress LLC Sulfo-cyanine7 succinimidyl ester (triethylamine) | 477908-53-5 | 95.7% | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD0824A10MG
Encompass_Preferred
CY7-SE triethylamine (Sulfo-Cyanine7 succinimidyl ester) is a near-infrared NHS-ester fluorescent labeling reagent supplied as a solid for conjugation to amines on proteins, antibodies, and small molecules. It exhibits excitation/emission at ~740/770 nm, has high extinction in the NIR range, and is supplied with documented purity and handling guidance.
Catalog No. 50-002-96897