Learn More
Medchemexpress LLC Sulfo DBCO-amine | 2028284-70-8 | MFCD30458046 | 99.0% | 427.47 g/mol | C21H21N3O5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1270565MG
Encompass_Preferred
Sulfo DBCO-amine is a water-soluble click-chemistry reagent containing a dibenzocyclooctyne (DBCO) moiety that enables strain-promoted alkyne-azide cycloaddition (SPAAC) for efficient, copper-free bioconjugation. It is commonly used as a linker or building block in PROTAC synthesis and other bioconjugation workflows, and is supplied in small quantities for research use.
  • Enables copper-free SPAAC bioconjugation.
  • Water-soluble sulfonate improves aqueous compatibility.
  • Contains a reactive amine for stable amide coupling.
  • High purity suitable for research-grade applications.
  • Available in small quantities for exploratory synthesis.
Catalog No. 50-002-63479