Learn More
Medchemexpress LLC Sulfo-EGS 100mg | 167410-92-6 | 616.48 g/mol | C18H20N2O18S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY117080100MG
Encompass_Preferred
Sulfo-EGS is a water-soluble, homobifunctional, amine-reactive crosslinker used to covalently link primary amines on proteins or peptides. It is commonly used to map protein-protein and protein-peptide interactions and to determine spatial proximity of amino acid residues within complexes.
  • Water-soluble crosslinker suitable for reactions in aqueous buffers.
  • Homobifunctional reagent that reacts with primary amines.
  • Enables mapping of protein-protein and protein-peptide interactions.
  • Useful for probing spatial proximity within protein complexes.
  • Solid form; store sealed at -20°C to maintain stability.
Catalog No. 50-003-08358