Learn More
Medchemexpress LLC Sulfo-EGS 10mg | 167410-92-6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11708010MG
Encompass_Preferred
Sulfo-EGS is a water-soluble, homobifunctional, amine-reactive crosslinker used to covalently link proximal primary amines in proteins and peptides to map interactions and probe complex topology. It is provided for research use in small milligram quantities.
  • Water-soluble homobifunctional amine-reactive crosslinker.
  • Targets primary amines for covalent crosslinking of proteins and peptides.
  • Facilitates mapping of protein-protein and protein-peptide interactions.
  • Suitable for small-scale, solution-phase crosslinking experiments.
  • Available in milligram pack sizes for research applications.
Catalog No. 50-003-07615