Learn More
Medchemexpress LLC Sulfo-EGS 25mg | 167410-92-6 | 616.48 | C18H20N2O18S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11708025MG
Encompass_Preferred
Sulfo-EGS is a water-soluble, homobifunctional, amine-reactive crosslinker used to probe protein-protein and protein-peptide interactions by forming stable covalent links between proximal lysine residues. It is soluble in water and DMSO and supplied for research use.
  • Water-soluble homobifunctional amine-reactive crosslinker.
  • Enables covalent crosslinking of proximal lysine residues to map interactions.
  • Soluble in water and DMSO for flexible experimental conditions.
  • Typical purity of 90.0% suitable for research applications.
  • Available in small pack sizes for bench-scale experiments.
Catalog No. 50-003-08359