Learn More
Medchemexpress LLC Sulfo-NHS-acetate sodium | 221222-61-3 | MFCD01861959 | 95.0% | 259.17 g·mol⁻¹ | C6H6NNaO7S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY140341A5MG
Encompass_Preferred
Sulfo-NHS-Acetate sodium is a water-soluble N-hydroxysuccinimide (NHS) ester used to acetylate primary amines and as a building block in PROTAC linker synthesis. Supplied as the sodium sulfonate salt for improved aqueous solubility, it enables efficient amine modification under mild, aqueous conditions for biochemical and chemical biology research.
  • Water-soluble sodium sulfonate form for improved handling in aqueous buffers.
  • Reacts with primary amines to introduce acetyl groups under mild conditions.
  • Suitable for bioconjugation, protein modification, and linker synthesis.
  • High purity appropriate for research applications.
  • Available in small pack sizes for low-scale synthesis and labeling studies.
Catalog No. 50-002-23131