Learn More
Abcam Sulfo-NHS (sulfo-N-hydro x ysuccinimide), Carbo x ylate-primary amine chemical modification reagent, 500MG
Supplier:  Abcam AB145608500MG
Encompass_Preferred
MW 217.13 Da, Purity >95%. Carboxylate-primary amine chemical modification reagent. Converts carboxyl groups to amine-reactive NHS esters for bioconjugation, crosslinking, labeling and immobilization methods. Increases the efficiency of EDC-mediated coupling reactions.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5116