Learn More
Aat Bioquest Sulfo-SMCC [4-N-Maleimidomethylcyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt] CAS 92921-24-9 (25 mg)
SDP
Supplier:  Aat Bioquest 4505
Encompass_Preferred
Sulfo-SMCC is the water-soluble version of SMCC. This catalog is for 25 mg.
Catalog No. 50-224-9394