Learn More
Medchemexpress LLC Sulfo-SMCC sodium | 92921-24-9 | 99.9% | 200 MG
SDP
Supplier:  Medchemexpress LLC HYD0975200MG
Encompass_Preferred
Sulfo-SMCC sodium is a commonly used hetero-bifunctional, noncleavable ADC crosslinker. It contains N-hydroxysuccinimide (NHS) ester and maleimide groups, which allow it to react with primary amines and sulfhydryl groups, respectively. This product is intended for research use only.
  • Reacts with primary amines and sulfhydryl groups
  • Suitable for ADC conjugation
  • Noncleavable linker
  • Used in various biological research applications
Catalog No. 50-004-09940