Learn More
Selleck Chemical LLC Sulfo-SMCC sodium S0566-50MG
SDP
Supplier:  Selleck Chemical LLC S056650MG
Encompass_Preferred
Sulfo-SMCC sodium is a hetero-bifunctional noncleavable ADC crosslinker which consists of N-hydroxysuccinimide (NHS) ester and maleimide groups to react with primary amines and sulfhydryl groups
Catalog No. 50-000-42561