Learn More
Medchemexpress LLC Sulfo-SPDB-DM4 | 1626359-59-8 | 99.8% | 1075.66 | C46H63ClN4O17S3 | 5 MG

Supplier: Medchemexpress LLC HY1011415MG
sulfo-SPDB-DM4 is a drug-linker conjugate for antibody-drug conjugate (ADC) research that delivers the maytansine-derived cytotoxin DM4 via a cleavable sulfo-SPDB linker. DM4 is an antitubulin maytansinoid that inhibits tubulin polymerization, causing mitotic arrest and apoptotic cell death. The product is supplied as a solid for use in ADC development and related in vitro studies.
- Delivers DM4, a maytansine-derived antitubulin payload, for ADC payload evaluation.
- Cleavable sulfo-SPDB linker enables intracellular release of the cytotoxin.
- High purity (≈99.8%) suitable for research applications.
- Supplied as a solid, white to yellow, shipped on dry ice.
- Soluble in DMSO at 50 MG/ML with ultrasonic assistance; hygroscopic DMSO affects solubility.
- Recommended storage at -80°C, protected from light, under nitrogen.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.