Learn More
Medchemexpress LLC Sulfo-SPP | 452072-27-4 | 97.1% | 420.48 g/mol | C14H16N2O7S3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12937710MG
Encompass_Preferred
Sulfo-SPP is a heterobifunctional, thiol-cleavable, membrane-impermeable crosslinker intended for research use. It is water-soluble and is used to selectively link thiol-containing biomolecules in aqueous environments.
  • Heterobifunctional crosslinker suitable for thiol conjugation.
  • Thiol-cleavable linkage enables controlled release of conjugated payloads.
  • Membrane-impermeable and water-soluble for aqueous bioconjugation.
  • Commonly used in antibody-drug conjugate and protein modification workflows.
  • Provided as a solid, white to off-white powder for easy handling and storage.
  • Stable under recommended storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-01997