Learn More
Medchemexpress LLC Sulfobetaine 10 | 15163-36-7 | 98.0% | 307.49 | 100 G
SDP
Supplier:  Medchemexpress LLC HY126377100G
Encompass_Preferred
Sulfobetaine 10 is a zwitterionic surfactant frequently utilized in biochemistry and biophysics. It serves to dissolve membrane proteins and other hydrophobic molecules in aqueous solutions. Its structure includes both positively and negatively charged groups, enabling it to maintain a neutral charge in solution and minimize protein denaturation. It is considered a mild, non-denaturing surfactant and is extensively employed in protein research. For research use only.
  • Zwitterionic surfactant
  • Utilized in biochemistry and biophysics
  • Dissolves membrane proteins and hydrophobic molecules
  • Maintains neutral charge in solution
  • Minimizes protein denaturation
  • Mild, non-denaturing surfactant
  • Widely employed in protein research
  • For research use only
Catalog No. 50-004-35831