Learn More
Medchemexpress LLC Sulfobetaine-16 100g
SDP
Supplier:  Medchemexpress LLC HYW099575100G
Encompass_Preferred
3-(Hexadecyldimethylammonio)propane-1-sulfonate is an organic compound belonging to the class of sulfonates It is a quaternary ammonium surfactant commonly used in various industrial and laboratory applications such as the production of detergents personal care products and pharmaceuticals 3-(Hexadecyldimethylammonio)propane-1-sulfonate is surface active and can be used as an emulsifier foaming agent and wetting agent In addition it is used as a solubilizer in protein purification and as a surfactant in chromatography
Catalog No. 50-002-06190