Learn More
TARGETMOL CHEMICALS INC Sulfometuron-methyl 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1976125MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Sulfometuron-methyl (DXP-5648) is a herbicide and also is an active antibacterial agent. It is an effective inhibitor of acetolactate synthase II and thus shows antibacterial properties. Purity 99.67%
Catalog No. 50-287-8344