Learn More
Cayman Chemical SulfosucInImIdyl MyrIstat 10mg
SDP
Supplier:  Cayman Chemical 1121210 MG
Encompass_Preferred
An inhibitor of fatty acid transport that has been used as an affinity label for certain membrane proteins that bind to fatty acids
Catalog No. 50-295-1342