Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfur, [3-cyano-5-[[[2,4-dimethyl-5-[6-(3-pyridinyl)-1H-imidazo[1,2-b]pyrazol-1-yl]... | 1448316-08-2 | 99.9% | 558.53 g·mol⁻¹ | C26H19F5N6OS | 1 ML
SDP

Supplier:  Medchemexpress LLC HY10075610MM 1ML

Encompass_Preferred

BAY-826 is a selective, potent small-molecule inhibitor of the TIE-2 kinase supplied for research use. It binds TIE-2 with high affinity (Kd ≈ 1.6 nM) and is provided as a 10 mM solution in DMSO (1 mL) or as solid quantities. Typical applications include biochemical kinase assays and cell-based studies of angiogenesis signaling pathways.

  • Selective TIE-2 inhibition with Kd ≈ 1.6 nM.
  • Available as 10 mM solution in DMSO (1 mL) and multiple solid sizes.
  • High purity suitable for research applications.
  • Molecular formula C26H19F5N6OS; molecular weight 558.53 g·mol⁻1.
  • Soluble in DMSO for convenient assay preparation.
  • Suitable for biochemical and cell-based assays targeting TIE-2 signaling.

Catalog No. 50-002-12005


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.