Learn More
Revvity Health Sciences Inc Sulfur-35 Radionuclide, 2mCi (74MBq), Specific Activity: 1050-1600Ci (38.8-59.2TBq)/mmol, Sulfuric Acid in 1mL Water
Supplier:  Revvity Health Sciences Inc NEX042002MC
Encompass
Sulfur-35 Radionuclide, 2mCi (74MBq), Specific Activity: 1050-1600Ci (38.8-59.2TBq)/mmol, Sulfuric Acid in 1mL Water
Catalog No. 50-281-1012