Learn More
Medchemexpress LLC Sulfur, [4-[[2-(1,1-difluoroethyl)-5-methyl[1,2,4]triazolo[1,5-a]pyrimidin-7-yl)amino]phenyl] | 1282041-94-4 | 99.9% | 415.33 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY100184
Encompass_Preferred
DSM265 is a long-duration inhibitor of P. falciparum dihydroorotate dehydrogenase (PfDHODH) with an IC50 of 8.9 nM. It can also inhibit the growth of Pf3D7 parasites with an EC50 of 4.3 nM.
  • Long-duration inhibitor of P. falciparum dihydroorotate dehydrogenase (PfDHODH).
  • Potent in vivo antimalarial activity with 90% effective dose (ED90) of 3 mg/kg per day in NOD-scid IL-2Rγnull (NSG) mice.
  • Moderate terminal elimination half-life (t1/2) of 2-4 hours in mice.
  • Appears as a solid, white to off-white substance.
Catalog No. 50-003-47080