Learn More
LAB ALLEY Lab Alley Sulfuric Acid 5% Solution
SDP
Supplier:  LAB ALLEY SUAL55GAL
Encompass
This chemical compound is categorized as an inorganic acid with a very low pH which requires precaution before using it as well as safe handling. Ideally it should be physically stored away from materials such as water organic materials metals amines bases and excess heat since it can generate undesirable reactions. A 5% sulfuric acid solution is a relatively dilute form of sulfuric acid which is commonly known as battery acid. Due to its lower concentration it is used in various applications where a milder acidic solution is required.
Catalog No. NC2965786