Learn More
Medchemexpress LLC Sulfuric acid ammonium salt (1:2) | 7783-20-2 | 99.3% | 132.14 | 100 G
SDP
Supplier:  Medchemexpress LLC 100GHYY0261A
Encompass_Preferred
Ammonium sulphate is an inorganic sulfate salt primarily used in molecular biology for applications such as precipitating proteins, separating antibodies, and enhancing antigen-antibody reactions. It also functions as a nitrogen source in plant fertilizers and has been observed to impact freshwater snails and mosquito larvae in rice paddies, as well as inhibit glucose conversion to ascorbic acid in plants. For research use only.
  • Used for molecular biology applications
  • Precipitates proteins
  • Separates antibodies
  • Enhances antigen-antibody reactions
  • Serves as a nitrogen source in plant fertilizers
  • Exhibits developmental toxicity to freshwater snails
  • Increases *Anopheles africanus* and *Culex* mosquito larvae populations
  • Inhibits the conversion of glucose to ascorbic acid in plants
Catalog No. 50-003-64939