Learn More
Medchemexpress LLC Sulfuric acid potassium salt (1:2) | 7778-80-5 | 99.2% | 174.26 | 500 G

Supplier: Medchemexpress LLC 500GHYY1083H
Potassium sulfate is a biological material or organic compound widely used in life science research. It can be used in biochemical tests for serum proteins, the determination of blood potassium and other potassium salts, and the preparation of protein capillary electrophoresis buffers.
- Used in biochemical tests for serum proteins
- Used for determination of blood potassium and other potassium salts
- Used in preparation of protein capillary electrophoresis buffers
- Widely used in life science research
- Molecular formula: K2SO4
- Appearance: Solid
- Purity: 99.2%
- CAS number: 7778-80-5
- Molecular weight: 174.26
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.