Learn More
Cayman Chemical SulIndac sulfon 5mg
SDP
Supplier:  Cayman Chemical 168055 MG
Encompass_Preferred
An oxidized metabolite of sulindac that is produced in many mammals but minimally in mice and rats; generally considered to be inactive against COX enzymes, although it can reduce azoxymethane-induced colon cancer in rats; inhibits aldose reductase (IC50 = 367 nM) in vitro
Catalog No. 50-296-8478