Learn More
Selleck Chemical LLC Suloctidil S5271-1g
SDP
Supplier:  Selleck Chemical LLC S52711G
Encompass_Preferred
Suloctidil a sulfur-containing aminoalcohol showing to inhibit human platelet aggregation is a peripheral vasodilator and a potential antifungal agent
Catalog No. 50-311-2208