Learn More
Medchemexpress LLC Sulopenem etzadroxil | 1000296-70-7 | 99.8% | 477.62 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10975410MG
Encompass_Preferred
Sulopenem etzadroxil is an orally active prodrug of the antibiotic Sulopenem. It demonstrates in vivo activity against *Bacillus anthracis* in a murine model, with survival rates comparable to those of the Ciprofloxacin-positive control. This makes it suitable for research in infection and antibacterial studies.
  • Orally active prodrug of Sulopenem
  • Active against *Bacillus anthracis*
  • Comparable survival rates to Ciprofloxacin in murine models
  • White to light yellow solid appearance
Catalog No. 50-003-45404