Learn More
Medchemexpress LLC SULT1A3 Human N-Hi 100ug
SDP
Supplier:  Medchemexpress LLC HYP71341A100UG
Encompass_Preferred
Recombinant human SULT1A3 protein expressed in Escherichia coli and purified with an N-terminal 6xHis tag. Supplied lyophilized for research use, the enzyme catalyzes sulfate conjugation of phenolic monoamines using PAPS and is suitable for biochemical assays, inhibitor screening, and antibody production. The preparation offers high purity and is formulated with stabilizers to support stability during storage and handling.
  • Supplied as 100 μg lyophilized material.
  • N-terminal His tag facilitates affinity purification and detection.
  • High purity (>95%) for reliable biochemical assays.
  • Full-length human sequence (295 aa) and ~35 kDa molecular weight.
  • Formulated with stabilizers for improved shelf life.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O.
  • Store at -20°C; aliquot after reconstitution for long-term use.
Catalog No. 50-001-96836