Learn More
Selleck Chemical LLC Sultamicillin S5411-1g
SDP
Supplier:  Selleck Chemical LLC S54111G
Encompass_Preferred
Sultamicillin is a newly developed antibiotic in which ampicillin and the -lactamase inhibitor sulbactam are linked as an ester
Catalog No. 50-310-8591