Learn More
Medchemexpress LLC Sultamicillin (tosylate) | 83105-70-8 | 1 G
SDP
Supplier:  Medchemexpress LLC HYN71111G
Encompass_Preferred
Sultamicillin tosylate is a broad-spectrum and orally active beta-lactamase inhibitor with antibacterial activity. It is an antibiotic that demonstrates effectiveness against various bacterial infections, including MRSA, in vitro.
  • Broad-spectrum beta-lactamase inhibitor
  • Displays antibacterial activity against MRSA (MIC value of 25 μg/mL)
  • Orally active, hydrolyzes into Sulbactam and Ampicillin upon absorption
  • Enhances bacterial clearance and improves survival in experimental polymicrobial sepsis in mice when combined with immunosuppression agents
  • Soluble in DMSO at 125 mg/mL
Catalog No. 50-004-18293