Learn More
Medchemexpress LLC Sultiame | 61-56-3 | MFCD00865304 | 98.9% | 290.4 g/mol | C10H14N2O4S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10831625MG
Encompass_Preferred
Sultiame is a sulfonamide carbonic anhydrase inhibitor used clinically as an antiepileptic and supplied for research and analytical applications. It is a stable, high-purity solid suitable for biochemical, pharmacological, and analytical studies.
  • Acts as a carbonic anhydrase inhibitor, modulating neuronal excitability.
  • Widely used as an antiepileptic agent in research contexts.
  • Supplied as a high-purity solid (listed purity ~98.9%).
  • Available in small research quantities for analytical use.
  • Includes supporting documentation such as SDS, COA, and datasheet.
Catalog No. 50-002-42692