Learn More
Medchemexpress LLC Sumatriptan succinate | 103628-48-4 | 13900 | 99.5% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB012150MG
Encompass_Preferred
Sumatriptan succinate is an orally active 5-HT1 receptor agonist used for research related to migraine headaches. It acts on 5-HT1D, 5-HT1B, and 5-HT1F receptors with specific IC50 values.
  • Orally active 5-HT1 receptor agonist
  • Used for migraine headache research
  • Exhibits IC50 values of 7.3 nM for 5-HT1D, 9.3 nM for 5-HT1B, and 17.8 nM for 5-HT1F receptors
  • Suitable for laboratory applications
Catalog No. 50-004-50863