Learn More
Medchemexpress LLC Sumatriptan succinate | 103628-48-4 | 99.5% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0121
Encompass_Preferred
Sumatriptan succinate is an orally active 5-HT1 receptor agonist, with IC50 values of 7.3 nM, 9.3 nM, and 17.8 nM for 5-HT1D, 5-HT1B, and 5-HT1F receptors, respectively. This compound is suitable for migraine headache research and has been shown to reverse nitroglycerin-induced thermal hypersensitivity in mice.
  • Orally active 5-HT1 receptor agonist
  • Suitable for migraine headache research
  • Reverses nitroglycerin-induced thermal hypersensitivity in mice
  • Exhibits IC50 values of 7.3 nM for 5-HT1D receptors
  • Exhibits IC50 values of 9.3 nM for 5-HT1B receptors
  • Exhibits IC50 values of 17.8 nM for 5-HT1F receptors
Catalog No. 50-004-06815