Learn More
Medchemexpress LLC Sumatriptan succinate | 103628-48-4 | 99.9% | 413.49 g/mol | C18H27N3O6S | 10 MG

Supplier: Medchemexpress LLC HYB012110MG
Sumatriptan succinate is an orally active 5-HT1 receptor agonist commonly used in migraine and serotonin receptor research. The compound is supplied as a white to off-white solid with high purity and useful aqueous solubility, and it is intended for analytical and pharmacological studies.
- High purity: 99.89%.
- Molecular weight: 413.49 g/mol.
- Chemical formula: C18H27N3O6S.
- Cas number: 103628-48-4.
- Appearance: white to off-white solid.
- Solubility: H2O ≥ 100 mg/mL; DMSO 50 mg/mL (may require ultrasonic).
- Storage: solid at 4°C; in solvent -80°C up to 6 months, -20°C up to 1 month.
- Applications: migraine and 5-HT1 receptor pharmacology research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.