Learn More
LifeSensors SUMO Protease 2 5000U
SDP
Supplier:  LifeSensors SP40205000
Encompass_Preferred
For removal of SUMOpro3 tags from fusion proteins expressed from the following SUMOpro3 expression systems: (Cat.#s: 1010A, 1016, 1010K, 1011, 2151). SUMO Protease 2, a highly active and robust recombinant protease, cleaves hSUMO3 from recombinant fusion proteins. Unlike thrombin, EK, or TEV protease, which recognize short, linear sequences, SUMO Protease 2 recognizes the tertiary structure of huSUMO3. As a result, SUMO Protease 2 will not cleave within the fused protein of interest. By purchasing this product, the purchaser agrees to comply with the terms of our Limited Use Label License.
Catalog No. NC2285778
Add to Cart
Request a Quote