Learn More
LifeSensors SUMO3-CHOP2 Reporter DeSUMOylation Assay
SDP
Supplier:  LifeSensors PR11020375
Encompass_Preferred
The SUMO3-CHOP2-Reporter DeSUMOylation Assay consists of human SUMO3 fused to a reporter enzyme, as well as a separate substrate for the reporter enzyme. When fused to SUMO3, the reporter is rendered catalytically inactive. Following cleavage of the SUMO3-reporter system by the isopeptidase, free (and now active) reporter subsequently acts upon its substrate. Thus, in this coupled assay, the signal generated by cleavage of the reporter enzyme’s substrate is a quantitative measure of isopeptidase activity. This product is protected by one or more US or Foreign patents.ContentsSUMO3-CHOP2-Reporter System: 1 x 375μL (4μM)SENP2 core (Control Isopeptidase): 2 x 10μL (2μM)Reporter Substrate 2: 1 x 30μL (25μM)Control Fluorophore 2 :1 x 20μL (1mM)
Catalog No. NC2956450