Learn More
Waters Corp Waters Column, C18, 100Å, 3.5 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 186002533
Encompass_Preferred
This C18 column is designed for general-purpose method development at low pH, providing high retention for basic compounds. It offers high-resolution, quick separation, and low-back pressure, suitable for various mass spectrometry applications. Manufactured in a cGMP, ISO 9001-certified laboratory, each batch is tested for repeatable performance.
  • High-resolution, quick, and low-back pressure.
  • Strong peaks, high peak capacity, outstanding sensitivity, and negligible bleed.
  • Superior low pH stability extends lifetime.
  • Provides symmetrical peaks for enhanced resolution and quantification.
Catalog No. 50-818-624