Learn More
Waters Corp Waters SunFire C18 Column, 100Å, 2.5 μm, 4.6 mm X 50 mm
Supplier:  Waters Corp 186003417
Encompass_Preferred
These C18 columns are designed for general purpose method development at low pH, featuring high retention for basic compounds and suitability for purification and impurity assays.
  • High retention for basic compounds; ideal for purification and impurity profile assays.
  • Superior low pH stability extends column lifetime.
  • Patented synthesis and bonding increase efficiency and sensitivity.
  • Effective design eliminates method reoptimization.
  • Comparable performance for scaling up analytical to preparative columns.
  • Achieves sharp peaks, outstanding sensitivity, and minimal bleed in MS applications.
  • Superior resolution, speed, and low back pressure reduce cost and analysis time.
  • Produces symmetrical peaks for better quantification and resolution of substances across pH ranges.
Catalog No. 50-781-754