Learn More
Waters Corp Waters Column, C18, 100Å, 3.5 μm, 2.1 × 150 mm
Supplier: Waters Corp 186002535
SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds and is ideally suited for purification and impurity profile assays.
- Features C18 chemistry and reversed phase separation for method development at low pH.
- Constructed with a silica particle substrate with low silanol activity and spherical particle shape.
- Supports a pH range of 2 to 8 and operates up to a maximum pressure of 6000 psi (415 bar).
- Endcapped for enhanced performance.
- Particle size is 3.5 μm and pore size is 100 Å.
- Suitable for HPLC systems, meeting USP classification L1.
- Inner diameter measures 2.1 mm and column length is 150 mm.
- Features a carbon load of 16 %, with one unit included per package.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.