Learn More
Waters Corp Waters Column, C18, 5 μm, 3 mm × 250 mm
Supplier:  Waters Corp 186002548
Encompass_Preferred
C18 columns are designed for general-purpose method development at low pH. The C18 ligand provides high retention, especially for basic compounds, making it suitable for purification and impurity profile assays.
  • Offers high-resolution, fast performance, and low-back pressure.
  • Features excellent sensitivity, high peak capacity, and minimal bleed, resulting in sharp peaks.
  • Versatile for various mass spectrometry applications.
  • Low pH stability extends its lifetime compared to some traditional silica-based HPLC columns.
  • Manufactured using patented, advanced silica synthesis, bonding, end-capping, and packing methods.
  • Contributes to improved peak shape performance and symmetrical peaks for better resolution and quantitation of acidic, neutral, and basic substances at low and intermediate pH ranges.
Catalog No. 50-781-188