Learn More
Waters Corp OBD Prep Column, C18, 100Å, 10 μm, 10 mm x 50 mm
Supplier: Waters Corp 186008208
SunFire C18 columns are designed for general purpose method development in low pH conditions, featuring a highly retentive C18 ligand. They are suitable for separating basic compounds and are ideally utilized in purification and impurity profile assays.
- Chemistry: C18
- Separation mode: Reversed phase
- Particle substrate: Silica
- pH range: 2 (min) to 8 (max)
- Temperature limits: 45 °C
- Maximum pressure: 6000 psi (415 bar)
- Endcapped: Yes
- Silanol activity: Low
- Particle shape: Spherical
- Particle size: 10 μm
- Pore size: 100 Å
- Format: OBD prep column
- Surface area: 340
- System: HPLC preparative
- Technique: LC/MS, LC
- USP classification: L1
- Inner diameter: 10 mm
- Length: 50 mm
- Carbon load: 16 %
- Product type: Preparative columns
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.