Learn More
Waters Corp Preparative Column, C18, 10 μm, 10 x 150 mm
Supplier:  Waters Corp 186008156
Encompass_Preferred
This SunFire C18 OBD Preparative Column features a 10 μm particle size and dimensions of 10 mm x 150 mm. It is designed for preparative chromatography applications.
  • Uniform, optimum packed bed density.
  • Direct scalability from analytical to preparative.
  • Extended column lifetime.
  • Consistent column to column performance.
  • Good resolution, peak shape, and mass loading for preparative chromatography results.
  • Maximize recoveries with robust packings designed for isolation of targeted small molecules and biomolecules.
  • Decrease risk of additional preparative method development.
Catalog No. 50-239-2483